missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ Split Straight-Through GC Inlet Liners
Supplied in blister packs to prevent contamination
Brand: SGE™ 092007
Code : 13
Additional Details : Weight : 0.50000kg
Description
- For various GC instruments
- All liners are deactivated using high temperature (420°C) gas phase deactivation
- Reduced active sites mean no unwanted reaction in samples
Specifications
GC Split Straight-through Liner | |
7.85 cm | |
0.63 cm |
5/Pk. | |
0.4 cm | |
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177 |