Learn More
Trajan™ SilTite™ FingerTite Ferrule System Adapters
For gas chromatography systems
Brand: Trajan™ 0736103
Additional Details : Weight : 0.20000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Adapter | |
Two Graphite Vespel FID Ferrules | |
Agilent FID adapter | |
Agilent FID |