Learn More
Trajan™ SilTite™ GC Ferrule Kit for Shimadzu Instruments
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan™ 073352
Code : 13
Additional Details : Weight : 1.00000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Shimadzu Injectors and Non-MS Detectors | |
GC Ferrule and Nut Kit | |
0.45 to 0.53 mm |
SilTite Metal | |
10 ferrules, 2 nuts |