Learn More
Trajan™ Replacement Plungers for eVol and eVol NMR Syringes
SGE syringe replacement plungers for gas tight syringes
£51.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12967596
|
Trajan™
2910380 |
eVol NMR Syringe (5μL) |
£51.00
Each |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12977596
|
Trajan™
2910382 |
eVol NMR Syringe (50μL) |
£51.00
Each |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12997596
|
Trajan™
2910384 |
eVol NMR Syringe (500μL) |
£51.00
Each |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Replacement Plunger |