Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Brand:  Trajan™ 031856
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15268563
  • £48.10 / Each
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
PTFE (Tip)
IN VITRO use only. For lab analyzers only. Not for human use.
Replacement Plunger, P5MD-GT
Syringe Needles (5 mL), Gas Tight Syringe