Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Brand: Trajan™ 031821
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- For use with SGE gas tight syringes.
- Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
PTFE (Tip) | |
IN VITRO use only. For lab analyzers only. Not for human use. | |
Replacement Plunger, P50F-C/T-GT | |
Syringe Needles (50μL), Fixed Needle Gas Tight Syringe |