Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Brand:  Trajan™ 0318120
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15258533
  • £48.10 / Pack of 2
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
  • For use with SGE gas tight syringes.
  • Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
PTFE (Tip)
IN VITRO use only. For lab analyzers only. Not for human use.
Replacement Plunger, P10F-C/T-GT
Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe