missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Replacement Needles for 25 to 500μL Removable Needle Syringes

Replacement Needles for 25 to 500uL Syringes for use in Chromatography

Brand:  Trajan™ 038038

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288593

  • £36.00 / Pack of 2
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Replacement Needles for 25 to 500uL Syringes for use in Chromatography

  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications

Specifications

Stainless Steel
0.009 in.
0.24 mm
Chromatography
Replacement Needle
Cone
23 G
0.024 in.
0.63 mm
7 in.
2/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.