missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Replacement Needles for 10μL Syringes

Made of Stainless steel

Brand:  SGE™ 037250

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10749891

  • £29.00 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Made of Stainless steel
Specifications

Specifications

22 ga.
0.72mm
Stainless steel
LC
0.17mm
For 10μL syringes
51mm
Replacement Needle
Videos
Documentation