missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ Replacement Needles for 0.5 - 1μL Syringes
For use with 0.5 to 1μL syringes
Brand: SGE™ 033630
Code : 13
Additional Details : Weight : 0.50000kg
Description
For removable-needle syringes or additional replacements, contact your Fisher Scientific Customer Service Representative.
Specifications
Stainless Steel | |
0.003 in. | |
0.1 mm | |
2.7 in. | |
Cone |
26 G | |
0.01 in. | |
0.47 mm | |
Replacement Needle | |
On-column Syringe |