Learn More
Trajan™ Replacement Kit for Plunger-in-Needle Syringes
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
Brand:  Trajan™ 034059
Code : 13
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10656601
  • £47.60 / Each
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched. Needles and plungers must be replaced as a set
Specifications
Specifications
23 G
0.63 mm
1 μL
Syringes (1μL)
0.63 mm
Replacement Needle Kit
Cone