missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Replacement Kit for Plunger-in-Needle Syringes
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
Brand: Trajan Scientific and Medical™ 034610
Code : 13
Additional Details : Weight : 0.50000kg
Description
- High precision and accuracy for small volume injections
- Replacement plunger/needle kits
- Kits are matched. Needles and plungers must be replaced as a set
Specifications
26 ga. | |
0.47mm | |
1μL Syringe with 8mm OD Barrel | |
Stainless steel | |
Stainless steel | |
Replacement needle and plunger: NP1B-7/0.47 |
0.47mm | |
1μL | |
Matched plunger and needle | |
70mm | |
Cone |