missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ PerkinElmer AutoSystem and Clarus 500 Liners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 092098

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12964031

  • £131.00 / Pack of 5


Estimated Shipment
18-05-2018
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with PerkinElmer instruments
Specifications

Specifications

Autosystem XL (PSS)
5/Pk.
2mm
86.2mm
AutoSystem and Clarus 500 liner
Split/Splitless
4mm
Videos