missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Manual Syringes For Rheodyne™ and Valco™ Valves

The injection size sets the injection volume

Brand:  SGE™ 002301

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10617171

  • £40.80 / Each


Estimated Shipment
09-02-2018
Add to basket

Specifications

Specifications

Fixed
Rheodyne™ and Valco™ injection valves
10F-LC
Each
22 ga.
0.71mm
50.8mm
LC
Manual Syringe
10μL
Videos
Documentation