Learn More
Trajan™ HT5 Aluminum-Clad Capillary Columns
Unique, high temperature phase suited for simulated distillation and other petroleum applications. Trajan Scientific and Medical™ HT5 Aluminum-Clad Capillary Columns are constructed from 5% Phenyl (equiv.) Polycarborane-siloxane.
Brand:  Trajan™ 054661
Code : 13
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10035440
  • £256.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Ultra High temperature column range
  • Unique phase - no equivalent phases
  • Ideal for simulated distillation applications (petroleum industry)
Specifications
Specifications
General Hydrocarbon profiles, pesticides, herbicides analysis
10 to 460/480°C
6m
5% Phenyl (equiv.) Polycarborane Siloxane
0.53mm
0.1μm
0.1μm
GC
SGE™ HT5
Videos