missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Guided Plunger Syringes

Extended barrel guides plunger during injection. SGE™ Guided Plunger Syringes are suitable for industrial environments.

Brand:  SGE™ 002450

Code : 13

Additional Details : Weight : 0.06016kg

 View more versions of this product

Product Code. 10452171

  • £98.00 / Each


In Stock 10
Add to basket

Specifications

Specifications

Removable
50mm
Bevel
Guided Plunger Syringe
10μL
0.47mm
10R-GP
Each
26 ga.
Videos
Documentation View accessory products