missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ BP10 (1701) GC Capillary Columns

Specifically designed for the separation of organochlorine pesticides listed in EPA methods 608 and 8081

Brand:  Trajan Scientific and Medical™ 054259

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12905541

  • £410.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Highly inert column
  • Low bleed
  • Can be solvent rinsed
  • Ideal for EPA methods 608 and 8081, pesticides, herbicides and pharmaceuticals
Specifications

Specifications

280/300°C
-20 to 280/300°C
30m
Forte™ BP10 (1701)
Fused Silica
Environmental Analyses (EPA Methods 608 and 8081), Pesticides/herbicides, Drugs of Abuse, Pharmaceuticals
G46
0.25μm
14% Cyanopropylphenyl Polysiloxane
Moderately Polar
0.25mm
Videos
Documentation