Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Brand:  Trajan™ 054064
Code : 13
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10742182
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
Van der Waals interaction
Phenols
G1, G2, G38
-60°C to +340/360°C
12 m
Non polar
0.32 mm
100% Dimethyl Polysiloxane
340/360°C
0.5 µm
Forte™ BP1
Fused Silica