missing translation for 'onlineSavingsMsg'
Learn More

SGE™ eVol™ Handheld Automated Analytical Syringe

Improves laboratory workflow

Brand:  SGE™ 2910322

Code : 13

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12937596

  • £223.00 / Pack of 3
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.

  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen

Applications:

  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution

Specifications

Specifications

25 G
0.2 mm
0.5 mm
eVol Analytical Syringe
50 μL
eVol Automated Syringe
0.007 in.
0.019 in.
1.96 in.
3/Pk.
Bevel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers