Learn More
Trajan™ CTC PAL Fixed Needle Syringe
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
Brand: Trajan™ 002983
Code : 16
Additional Details : Weight : 0.10000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
-
Specifications
Gas tight | |
0.72 mm | |
Fixed Needle Syringe | |
1 Ea. | |
Side Hole |
22 G | |
0.72 mm | |
10F-CTC-GT-5/0.72H | |
10 μL |