missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BP21 (FFAP) GC Capillary Columns

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

Brand:  Trajan™ 054468

Code : 13

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12975591

  • £565.00 / Each
Estimated Shipment: 03-05-2024
to see stock.
Add to basket

Description

Description

A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids

  • Nitroterephthalic acid modified PEG polar phase
  • Ideal for low molecular weight acids
  • Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications

Specifications

0.32 mm
Volatile Free Acids, Fatty Acid Methyl Esters, Alcohols, Aldehydes, Acrylates, Ketones
Acrylates Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones
35°C to 240/250°C
25 m
Polar
G25, G35
Polyethylene Glycol
0.25 μm
0.25 μm
BP21 (FFAP)
Fused Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers