missing translation for 'onlineSavingsMsg'
Learn More

SGE™ BP20 (WAX) Capillary Columns

Industry standard WAX columns

Brand:  SGE™ 054451

Code : 13

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12995581

  • £620.00 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  •  Very polar polyethylene glycol (PEG) phase
  • Cross-linked for stability and washing
  • Also ideal for xylene and aromatic isomer
  • 0.1 to 1μm film thickness: 20° to 260/280°C
Specifications

Specifications

0.53 mm
Ketones Analysis, Ketones, Aldehydes Analysis, Fragrance Analysis
1 μm
Polar
SGE™ BP20 (Wax)
1 μm
30 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers