missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ sFRP-1 Inhibitor

Brand:  Alfa Aesar™ J65910.MB

Code : NEW

Additional Details : CAS Number : 915754-88-0 Weight : 0.00025kg

 View more versions of this product

Product Code. 15497119

  • £364.32 / 25mg


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Description

Description

A diphenylsulfone-sulfonamide compound that counteracts the antagonizing effect of SARP-2/sFRP-1.
Specifications

Specifications

25mg
2° to 8°C
A diphenylsulfone-sulfonamide compound that counteracts the antagonizing effect of SARP-2/sFRP-1
C19H26N2O4S2
Secreted Frizzled Related Protein-1 Inhibitor; N-(3-(Dimethylamino)propyl)-2-ethyl-5-(phenylsulfonyl)benzenesulfonamide
Signal Transduction Reagent
Clear
Oil
915754-88-0
410.60g/mol
Light Sensitive
Videos
SDS