Learn More
Mettler Toledo™ SevenGo™ Portable Conductivity Meters
Brand:  Mettler Toledo™ 51302535
Code : 59
Additional Details : Weight : 0.33000kg
Product Code. 11997991
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
SevenGo Portable Conductivity Meter
Instrument only
0.01 to 1
0.01 mg/L to 300 g/L
± 0.5%
0.1 deg.C
1.5V
Automatic buffer recognition, temperature compensation and end point
SG3-B
0.10 uS/cm to 500 mS/cm
± 0.5%
0.01 to 1
0 to 80ppt
±0.2 deg.C
IP67
–5 to 105°C
Alternative Products