missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenGo™ Portable Conductivity Meters

Brand:  Mettler Toledo™ 51302571

Code : 04

Additional Details : Weight : 0.34000kg

Product Code. 10163412

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

All kits as standard equipment include: wrist strap, 4 x AA Batteries, operating instructions, test certificates and declaration of conformity.
  • Ergonomically designed and robust, with IP67 protection
  • Large easy to read displays (backlit on the professional versions)
  • Clearly defined, easy to read buttons (simple menu structure on professional versions), intuitive operation
  • Large built-in data memory for detailed GLP protocols
  • Automatic buffer recognition, temperature compensation and end point
  • Infrared interface for data transfer to printer or PC
Specifications

Specifications

SevenGo Portable Conductivity Meter
Electrode kit with InLab 738 (IP67, 1.8m cable)
± 0.5%
0 to 80ppt
±0.1 deg.C
IP67
–5 to 105°C
SG7-ELK
0.01 uS/cm to 1000 mS/cm
0.01 mg/L to 600 g/L
0.0 to 100Mohm.cm
1.5V
Automatic buffer recognition, temperature compensation and end point
Videos
Documentation