Mettler Toledo™ SevenGo™ Portable Conductivity Meters


Overview
Brand: Mettler Toledo™

Manufacturer Part Number: 51302573

UNSPSC: 41115609

Code: 04

Additional Details:
Additional Details: Weight: 0.34000kg



Product Code. 10519932

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


  • Description and Specification
  • Alternative Products
  • Documentation
  • Accessories and Spare Parts
Description and Specification

Specification

Range (Conductivity) 0.01 uS/cm to 1000 mS/cm
Range (Resistivity) 0.0 to 100Mohm.cm
Range (TDS) 0.01 mg/L to 600 g/L
Model SG7-FK
Range (Salinity) 0 to 80ppt
IP Rating IP67
Product Type SevenGo Portable Conductivity Meter
Accuracy (Temperature) ±0.1 deg.C
Temperature Range (Metric) –5 to 105°C
Battery Type 1.5V
For Use With (Application) Automatic buffer recognition, temperature compensation and end point
Includes Field kit with InLab 738 (IP67, 1.8m cable)
Accuracy (Conductivity) ± 0.5%

All kits as standard equipment include: wrist strap, 4 x AA Batteries, operating instructions, test certificates and declaration of conformity.
  • Ergonomically designed and robust, with IP67 protection
  • Large easy to read displays (backlit on the professional versions)
  • Clearly defined, easy to read buttons (simple menu structure on professional versions), intuitive operation
  • Large built-in data memory for detailed GLP protocols
  • Automatic buffer recognition, temperature compensation and end point
  • Infrared interface for data transfer to printer or PC

Alternative Products
Documentation Accessories and Spare Parts