missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenGo™ Portable Conductivity Meters

Brand:  Mettler Toledo™ 51302576

UNSPSC : 41115609

Code : 59

Additional Details : Weight : 2.00000kg

Product Code. 11918001

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications Description & Specifications

Specifications

SevenGo Portable Conductivity Meter
Instrument only
± 0.5%
0 to 80ppt
±0.1 deg.C
IP67
10m
SG7-B
0.01 uS/cm to 1000 mS/cm
0.01 mg/L to 600 g/L
0.0 to 100Mohm.cm
1.5V
Automatic buffer recognition, temperature compensation and end point
–5 to 105°C
Alternative Products