missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ S210 SevenCompact™ pH, mV/ORP-Bio
This well-designed meter is the perfect choice for a wide range of applications without overstretching your budget. Mettler Toledo™ S210 SevenCompact™ pH, mV/ORP-Bio can also measure ORP.
Brand: Mettler Toledo™ 30130864
Code : NEW
Additional Details : Weight : 2.84000kg
Description
- Includes electrode holder
- UPlace™ movement helps bring your electrode into the best position for your sample
- Fast measurements and reduced sample tipping risk
- Reduced sensor damage risk
- Choice of viewing all measurement information on the display or focusing on the essentials with the uFocus™ view
- Switching between the different display layouts requires only a single keypress
- Color display with well designed icons
- Menu settings in 10 languages
- Security and high reproducibility thanks to Intelligent Sensor Management (ISM™)
- Professional calibration support
- Comprehensive service package
- Integrated USB and RS232 interfaces
- Includes InLab™ Routine Pro-ISM, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21, and 10.0
Specifications
±0.2 mV | |
9 to 12 V | |
SevenCompact pH meter S210-Bio-Kit | |
Single-channel | |
-2000 to 2000.0 | |
-30.0°C to +130°C (MTC), -5.0°C to +130.0°C (ATC) | |
InLab™ Routine Pro-ISM, guide to pH measurements and 2 buffer sachets for pH 4.01, 7.00, 9.21, and 10.0 | |
1.000 measurements | |
9 to 12V/10W | |
0.1/1mV | |
9 to 12V, 10W |
±0.002 | |
Max. 5 points, 8 pre-defined and 1 user-defined buffer group | |
-2 to 20°C | |
TFT Color, 4.34 in. | |
-2 to 20 | |
Measurements of pH, mV/ORP | |
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
pH bench meter SC S210-Bio | |
pH/mV/ORP Meter | |
±0.1°C | |
9 to 12V |