Learn More
Fisher Chemical Seven components; Fluorides (F-) 20mg/L, Chlorides (Cl-) 100mg/L, Nitrites (NO2-) 100mg/L, Bromides (Br-) 100mg/L, Nitrates (NO3-) 100mg/L, Phosphates (PO43-) 200mg/L, Sulphates (SO42-) 100mg/L for Ion Chromatography H2O matrix
Brand:  Fisher Chemical CM5580
Seven components; Fluorides (F-) 20mg/L, Chlorides (Cl-) 100mg/L, Nitrites (NO2-) 100mg/L, Bromides (Br-) 100mg/L, Nitrates (NO3-) 100mg/L, Phosphates (PO43-) 200mg/L, Sulphates (SO42-) 100mg/L for Ion Chromatography H2O matrix
Code : 04
Additional Details : Weight : 0.05000kg
Product Code. 12964435