missing translation for 'onlineSavingsMsg'
Learn More

Sertraline hydrochloride, Tocris Bioscience™

5-HT reuptake inhibitor

Brand:  Tocris (Bio-Techne) 239550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431134

  • £655.62 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, orally active selective serotonin re-uptake inhibitor (SSRI); antidepressant.
Specifications

Chemical Identifiers

79559-97-0
342.688
sertraline hydrochloride, sertraline hcl, zoloft, gladem, serad, 1s,4s-4-3,4-dichlorophenyl-n-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride, atruline, tresleen, tatig
CHEBI:9124
CNC1CCC(C2=CC=CC=C12)C3=CC(=C(C=C3)Cl)Cl.Cl
C17H18Cl3N
BLFQGGGGFNSJKA-XHXSRVRCSA-N
63009
(1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride

Specifications

50mg
79559-97-0
sertraline hydrochloride, sertraline hcl, zoloft, gladem, serad, 1s,4s-4-3,4-dichlorophenyl-n-methyl-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride, atruline, tresleen, tatig
CNC1CCC(C2=CC=CC=C12)C3=CC(=C(C=C3)Cl)Cl.Cl
342.688
CHEBI:9124
>99%
Sertraline hydrochloride
C17H18Cl3N
BLFQGGGGFNSJKA-XHXSRVRCSA-N
(1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
63009
342.69
Videos
Documents