Learn More
SERTAD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554278
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPPALLPEKD SFSSALDEIE ELCPTSTSTE AATAATDSVK GTSSEAGTQK LDGPQESRAD DSKLMDSLPG NFEITTSTGF LTDLTLDDIL FADIDTSMYD FDPCTSSSGT ASKMAPVSAD DLLKTLAP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
SERTAD2 Polyclonal antibody specifically detects SERTAD2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SERTAD2 | |
Polyclonal | |
Unconjugated | |
SERTAD2 | |
AB041541; AU021878; Kiaa0127; MGC126688; MGC126690; mKIAA0127; MNCb-1504; PHD zinc finger- and bromodomain-interacting protein 2; Sei2; Sei-2; SERTA domain containing 2; SERTA domain-containing protein 2; SERTAD2; Transcriptional regulator interacting with the PHD-bromodomain 2; transcriptional regulator interacting with the PHS-bromodomain 2; TRIPBR2; TRIP-Br2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14140 | |
SERTAD2 | |
Recombinant protein corresponding to Human SERTAD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only