missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin I2 Mouse anti-Human, Clone: 203402, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Serpin I2 |
---|---|
Clone | 203402 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Serpin I2 |
Gene ID (Entrez) | 5276 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
Serpin I2 Monoclonal specifically detects Serpin I2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Serpin I2 | |
Mouse myeloma cell line NS0-derived recombinant human Serpin I2 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Serpin I2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Serpin A1, A3, A4, A5, C1, F2, I1, recombinant mouse (rm) Serpin D1, or rmPEDF is observed. | |
Unconjugated | |
Western Blot, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
203402 | |
5276 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
For Research Use Only