missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin F1/PEDF Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Serpin F1/PEDF |
---|---|
Clone | 199315 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Serpin F1/PEDF Monoclonal specifically detects Serpin F1/PEDF in Mouse samples. It is validated for Western Blot.Specifications
Serpin F1/PEDF | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P97298 | |
5176 | |
E. coli-derived recombinant mouse Serpin F1/PEDF Asp43-Thr417 Accession # P97298 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Serpin F1/PEDF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Serpin A1, A3, A5, C1, or F1. |
199315 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cell proliferation-inducing gene 35 protein, EPC-1, EPC-1PIG35, PEDF, PEDFpigment epithelium-derived factor, pigment epithelium derived factor), member 1, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, Serpin F1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 1 | |
SERPINF1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only