missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin E2/PN1 Rat anti-Mouse, Clone: 244404, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Serpin E2/PN1 |
---|---|
Clone | 244404 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1 Ser20-Pro397 Accession # Q07235 |
Gene ID (Entrez) | 5270 |
Gene Accession No. | Q07235 |
Description
Serpin E2/PN1 Monoclonal specifically detects Serpin E2/PN1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Serpin E2/PN1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin E2/PN1 Ser20-Pro397 Accession # Q07235 | |
Q07235 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
244404 | |
5270 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Serpin A1, recombinant mouse (rm) Serpin D1, or rmSerpin F2 is observed. | |
Unconjugated | |
Western Blot, Immunoprecipitation | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only