Learn More
Serpin E2/PN1 Mouse anti-Human, Clone: 449888, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2980
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439825
  • £288.60 / 100µg
Estimated Shipment date
from Supplier 30-06-2020
Description
Description
Serpin E2/PN1 Monoclonal antibody MAB2980 Specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Serpin E2/PN1
449888
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINE2
Mouse myeloma cell line NS0-derived recombinant human Serpin E2/PN1 Met1-Pro397 Accession # NP_001130000
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5270
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_001130000
GDN, GdNPF, GDNserine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 2, glia-derived nexin, glial-derived neurite promoting factor, Peptidase inhibitor 7, PI-7, PI7DKFZp686A13110, PN-1, PN1nexin, PNI, Protease nexin 1, Protease nexin I, Proteinase Nexin 1, Serpin E2, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 2
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Serpin E2/PN1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Serpin A1, A3, A4, A5, A12, B6, B8, B9, C1, D1, F2, I1, recombinant mouse Serpin E2, or rhPEDF is observed.