missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Serpin D1/Heparin Cofactor II Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Serpin D1/Heparin Cofactor II |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.23 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Description
Serpin D1/Heparin Cofactor II Polyclonal specifically detects Serpin D1/Heparin Cofactor II in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Serpin D1/Heparin Cofactor II | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.23 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
HC2, HC2LS2, HCF2, HCF2clade D (heparin cofactor), member 1, HC-II, Heparin cofactor II, HLS2HCII, leuserpin 2, LS2, Protease inhibitor leuserpin-2, Serpin D1, serpin peptidase inhibitor, clade D (heparin cofactor), member 1 | |
SERPIND1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
P49182 | |
3053 | |
Mouse myeloma cell line NS0-derived recombinant mouse Serpin D1/Heparin Cofactor II Glu24-Ser478 Accession # P49182 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Serpin D1/Heparin Cofactor II in direct ELISAs and Western blots. |
For Research Use Only