missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A3/alpha 1-Antichymotrypsin Mouse anti-Human, Clone: 213907, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Serpin A3/alpha 1-Antichymotrypsin |
---|---|
Clone | 213907 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Serpin A3 aa 26-423 Accession # P01011 |
Gene ID (Entrez) | 12 |
Gene Accession No. | P01011 |
Description
Serpin A3/alpha 1-Antichymotrypsin Monoclonal specifically detects Serpin A3/alpha 1-Antichymotrypsin in Human samples. It is validated for Western Blot.Specifications
Serpin A3/alpha 1-Antichymotrypsin | |
Mouse myeloma cell line NS0-derived recombinant human Serpin A3 aa 26-423 Accession # P01011 | |
P01011 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
213907 | |
12 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects recombinant human (rh)Serpin A3 in direct ELISAs and Western blots. In these formats, this antibody shows 5 - 25% cross-reactivity with rhSerpins A1 and A5 but no cross-reactivity with rhSerpins C1, D1, or F2. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only