Learn More
National Diagnostics™ SequaGel™ XR Gel Solution
Brand:  National Diagnostics™ EC-842-450
Code : LE
Additional Details : Weight : 0.01000kg
Product Code. 12341459
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
National Diagnostics' SequaGel™ XR is specially formulated to produce greater resolution and more information - up to 500 DNA bands on a single-load gel.
SequaGel™ XR's optimised, proprietary formulation prevents aberrant band inversions and yields sharp, clear, highly resolved bands.Runs substantially faster than traditional gels, while actually improving resolution. These results are achieved in a comfortable time-tested format, providing full inter-laboratory reproducibility. By preventing band inversions, SequaGel™ XR bands are sharp and true. SequaGel™ XR provides more information by allowing greater separation between bands near the top of the gel. This effect, similar to a wedge gel, is achieved with standard spacers.The quality and consistency of SequaGel™ XR is shown by the uniform background obtained upon drying. Band edges are sharp. Even without fixing, the gel will dry without cracking or crazing. The result is a finely-rendered, highly detailed DNA gel.Shipped as a two component system consisting of the SequaGel™ XR monomer solution and SequaGel™ complete buffer. SequaGel™ complete buffer solution contains TBE and TEMED. No dilutions are necessary in this easy-to-use system.The user simply combines the two solutions, adds ammonium persulfate, and casts the gel. SequaGel™ XR is also available in the form of a 50% stock solution, SequaGel™ XR Concentrate.
SequaGel™ MD
  • For detection of minor mutational differences
  • Point mutation analysis/SSCP analysisSupplied as 2X stock solution
SequaGel™ XR
Consistently crystal clear gels
Stabilised for a two year shelf life
Specifications
Specifications
450mL
Videos
Documentation
Technical Documents