missing translation for 'onlineSavingsMsg'
Learn More

National Diagnostics™ SequaGel™ UreaGel™ Concentrate

Brand:  National Diagnostics™ EC-830-1

Code : LF

Additional Details : Weight : 0.01000kg


Product Code. 12351439

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The SequaGel™ sequencing system consists of SequaGel™ Concentrate, SequaGel™ Diluent, and SequaGel™ Buffer. This system provides a convenient, dependable means for researchers to prepare gels of varying percentage. With the SequaGel™ sequencing system the researcher readily prepares any commonly used gel formulation up to 20% monomer (19:1 Acrylamide: Bis-acrylamide). 1L bottle of SequaGel™ Concentrate contains 237.5g of acrylamide, 12. 5g of methylene bisacrylamide, and urea in a deionised aqueous solution. SequaGel™ Diluent is supplied in 450mL and 1L bottle containing urea in deionised water. SequaGel™ Buffer is supplied in 100mL and 200mL bottles containing 0.89M Tris-Borate-20mM EDTA buffer pH8.3 (10X TBE) and urea.
  • Casts 19:1 denaturing gels up to 20% monomer
  • Aldehyde free and ammonia free
  • Stabilised for long shelf life
  • Consistently crystal clear gels For DNA sequencing, manual.
Specifications

Specifications

Acrylamide, bisacrylamide, urea
1L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • UreaGel Concentrate¬†

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P306+P360-IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains¬†: Acrylamide

Documentation

Technical Documents