missing translation for 'onlineSavingsMsg'
Learn More

National Diagnostics™ SequaGel™ UreaGel™ Concentrate

Brand:  National Diagnostics™ EC-830-450

UNSPSC : 41105318

Code : LF

Additional Details : Weight : 0.01000kg

Product Code. 12361439

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The SequaGel™ sequencing system consists of SequaGel™ Concentrate, SequaGel™ Diluent, and SequaGel™ Buffer. This system provides a convenient, dependable means for researchers to prepare gels of varying percentage. With the SequaGel™ sequencing system the researcher readily prepares any commonly used gel formulation up to 20% monomer (19:1 Acrylamide: Bis-acrylamide). 1L bottle of SequaGel™ Concentrate contains 237.5g of acrylamide, 12. 5g of methylene bisacrylamide, and urea in a deionised aqueous solution. SequaGel™ Diluent is supplied in 450mL and 1L bottle containing urea in deionised water. SequaGel™ Buffer is supplied in 100mL and 200mL bottles containing 0.89M Tris-Borate-20mM EDTA buffer pH8.3 (10X TBE) and urea.
  • Casts 19:1 denaturing gels up to 20% monomer
  • Aldehyde free and ammonia free
  • Stabilised for long shelf life
  • Consistently crystal clear gels For DNA sequencing, manual.
Specifications

Specifications

Urea, Acrylamide, Bis-acrylamide
DNA sequencing
450mL
Documentation

Technical Documents