Learn More
National Diagnostics™ SequaGel™ UreaGel™ 6
Brand:  National Diagnostics™ EC-836-450
Code : LF
Additional Details : Weight : 0.01000kg
Product Code. 12381449
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
SequaGels™ 4, 4.25, 4.75, 6 and 8 each consists of SequaGel™ monomer solution and SequaGel™ complete buffer solution. The respective SequaGel™ monomer solution contains urea, as well as acrylamide and bis-acrylamide in the standard 19:1 ratio. SequaGel™ complete buffer solution contains TBE and TEMED. Upon combining these two solutions, the researcher adds ammonium persulfate to form a crystal clear electrophoresis matrix with corresponding percent acrylamide (4, 4.25, 4.75, 6, or 8) containing 1X TBE (89mM Tris base, 89mM boric acid, 2mM EDTA, pH8.3) and 6M urea. Easy to use and reliable.
  • Ready-to-use 19:1 denaturing DNA gel solutions
  • Aldehyde and ammonia free
  • Consistently crystal clear gels
  • Twelve month shelf life at room temperature For DNA sequencing.
Specifications
Specifications
Urea, Acrylamide, Bis-acrylamide, TBE, TEMED, Ammonium Persulfate
Denaturing DNA
450mL
Free
Free
Videos
Documentation
Technical Documents
View accessory products