Learn More
Septin-7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556181
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VNIIPLIAKA DTLTPEECQQ FKKQIMKEIQ EHKIKIYEFP ETDDEEENKL VKKIKDRLPL AVVGSNTIIE VNGKRVRGRQ YPW Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Septin-7 Polyclonal antibody specifically detects Septin-7 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Septin-7 | |
Polyclonal | |
Unconjugated | |
SEPTIN7 | |
7-Sep; Cdc10; CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC10 (cell division cycle 10, S.cerevisiae, homolog); CDC10 protein homolog; CDC3; cell division cycle 10 homolog; E430034N22; NBLA02942; SEPT7; SEPT7A; septin 7; septin 7 variant 4; Septin7; septin-7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
235072, 64551, 989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O55131, Q16181, Q9WVC0 | |
SEPTIN7 | |
Recombinant protein corresponding to Human Septin-7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only