missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Septa for 0.2 mL MicroAmp™ tubes (for the 310 Genetic Analyzer)
These are septa covers for 0.2mL sample tubes, for use with the 310 Genetic Analyzer.
Brand: Applied Biosystems™ 4305547
Additional Details : Weight : 0.01000kg
Specifications
Each package contains 480 septa. | |
Sequencing | |
Septa | |
Room Temperature |
310 Genetic Analyzer | |
MicroAmp™ | |
480 Each | |
Disposable Tube, Conical Tube, Assay Tube |
For research use only