missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Separating or resolving buffer (4X)

Brand:  Alfa Aesar™ J60181.K2

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15402999

  • £65.50 / 1 L

Estimated Shipment: 31-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4X
Liquid
Ambient temperatures
1L
Videos
SDS Documents
Certificates