Learn More
Ansell™ Sensilite™ 48-101 Series Black Polyurethane Light Weight Gloves
Designed for light applications that require good levels of comfort and abrasion resistance.
Brand: Ansell™ 48101110
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Industrial gloves
- Abrasion resistance for extended wear life
- Thin liner and coating offering great dexterity and flexibility
- Sensilite 48-100 allows extreme tactility where precision is required
- Comfort and protection for lightweight applications
- Offers precise handling of small objects in dry or light oily environments
- Provides enhanced safety, dexterity and durability
Applications:
- Assembly and disassembly of equipment
- Chassis assembly, body trimming
- Equipment repair and maintenance
- Loading and unloading rough parts or boxes
- Picking parts and accessories
- Production line support and maintenance
Specifications
Polyurethane | |
Knitwrist | |
Coated-palm | |
48-101 | |
12 Pairs | |
Standard CATEGORY II EN 388 : 4131 |
Nylon | |
Nylon | |
Abrasion resistance | |
11 | |
Black |