Learn More
Semiquantitative Standard 1, Specpure™, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 047180.AP
7459.03 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00050kg Transport : UN number : 3093 Chem class : 8 Pack group : II
Description
Semiquantitative Standard 1, Specpure™ is used as a standard for inductively coupled plasma and direct current plasma (DCP) emission spectrometry for the analysis of trace elements in biomedical and environmental samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Semiquantitative Standard 1 | |
MFCD02091742 | |
Specpure™ | |
Liquid |
500 mL | |
UN3093 | |
Miscible with water |
RUO – Research Use Only