missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 6A Mouse anti-Human, Clone: 169203, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Semaphorin 6A |
---|---|
Clone | 169203 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A Gly19-Thr649 Accession # Q9H2E6 |
Gene ID (Entrez) | 57556 |
Gene Accession No. | Q9H2E6 |
Description
Semaphorin 6A Monoclonal specifically detects Semaphorin 6A in Human samples. It is validated for Western Blot.Specifications
Semaphorin 6A | |
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A Gly19-Thr649 Accession # Q9H2E6 | |
Q9H2E6 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
169203 | |
57556 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human Semaphorin 6A in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) Semaphorin 6A is observed and no cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 3B, or rmSemaphorin 3F is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only