Learn More
SEMA4A Monoclonal Antibody (5E3), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-9753-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.
SEMA4A Monoclonal antibody specifically detects SEMA4A in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
SEMA4A | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q62178, Q9H3S1 | |
SEMA4A | |
Affinity chromatography | |
RUO | |
20351, 64218 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
5E3 | |
PerCP-eFluor 710 | |
SEMA4A | |
AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 | |
Mouse | |
25 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.