missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 3E Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Semaphorin 3E |
---|---|
Clone | 400520 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Semaphorin 3E Monoclonal specifically detects Semaphorin 3E in Human samples. It is validated for Western Blot.Specifications
Semaphorin 3E | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O15041 | |
9723 | |
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E Thr25-Ser775 (Arg557Ala and Arg560Ala) Accession # O15041 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Semaphorin 3E in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) Semaphorin 3A, 3B, 6A, 6B, 6C, 6D, 7A, rmSemaphorin 3C, 3E, or 3F is observed. |
400520 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
(semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E | |
SEMA3E | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only