Allsigns International™ Self-Adhesive Vinyl Hazard Labels


Overview
Brand: Allsigns International™

Manufacturer Part Number: DA1(S)

SELF ADHESIVE VINYL SIGN:- `FLAMMABLE LIQUID'102²M

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12355768

Quantity Price
1 £ 7.98 / Each
4 £ 6.39 / Each
Estimated Shipment
01-02-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Specific uses wherever hazardous materials are in use
Height (Metric) 102mm
Adhesive Type Self-adhesive
Quantity 1Pack
Type Hazard Label
Width (Metric) 102mm
Material Vinyl
Message Flammable Liquid
Length (Metric) 102mm

Sold as single labels, these hazard diamonds are ideal for specific uses wherever hazardous materials are in use. In self adhesive vinyl, dimensions 100mm x 100mm.

Documentation